Merck
  • Decreased Mitochondrial DNA Content Drives OXPHOS Dysregulation in Chromophobe Renal Cell Carcinoma.
Decreased Mitochondrial DNA Content Drives OXPHOS Dysregulation in Chromophobe Renal Cell Carcinoma.
Cancer research (2020-07-23)
Yi Xiao, Rosanna Clima, Jonas Busch, Anja Rabien, Ergin Kilic, Sonia L Villegas, Bernd Timmermann, Marcella Attimonelli, Klaus Jung, David Meierhofer
ABSTRACT
Chromophobe renal cell carcinoma (chRCC) and renal oncocytoma are closely related, rare kidney tumors. Mutations in complex I (CI)-encoding genes play an important role in dysfunction of the oxidative phosphorylation (OXPHOS) system in renal oncocytoma, but are less frequently observed in chRCC. As such, the relevance of OXPHOS status and role of CI mutations in chRCC remain unknown. To address this issue, we performed proteome and metabolome profiling as well as mitochondrial whole-exome sequencing to detect mitochondrial alterations in chRCC tissue specimens. Multiomic analysis revealed downregulation of electron transport chain (ETC) components in chRCC that differed from the expression profile in renal oncocytoma. A decrease in mitochondrial (mt)DNA content, rather than CI mutations, was the main cause for reduced OXPHOS in chRCC. There was a negative correlation between protein and transcript levels of nuclear DNA- but not mtDNA-encoded ETC complex subunits in chRCC. In addition, the reactive oxygen species scavenger glutathione (GSH) was upregulated in chRCC due to decreased expression of proteins involved in GSH degradation. These results demonstrate that distinct mechanisms of OXPHOS exist in chRCC and renal oncocytoma and that expression levels of ETC complex subunits can serve as a diagnostic marker for this rare malignancy. SIGNIFICANCE: These findings establish potential diagnostic markers to distinguish malignant chRCC from its highly similar but benign counterpart, renal oncocytoma.
Sigma-Aldrich
Anti-UQCRC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SDHB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
L-(−)-Glucose, ≥99%