
Plastic additive 15

Synonym(s):
Dioctadecyl disulfide
Empirical Formula (Hill Notation):
C36H74S2
CAS Number:
Molecular Weight:
571.10
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing