
Paraquat dichloride-(rings-d8) hydrate

Synonym(s):
1,1′-Dimethyl-4,4′-bipyridinium-2,2′,3,3′,5,5′,6,6′-d8 dichloride, Methyl viologen-d8 dichloride, Paraquat dichloride-d8
Empirical Formula (Hill Notation):
C12D8H6Cl2N2 · xH2O
Molecular Weight:
265.21 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing