
Lithium-6Li2 carbonate

Synonym(s):
Lithium carbonate-6Li2, Lithium-6Li2 carbonate
Linear Formula:
6Li2CO3
CAS Number:
Molecular Weight:
72.04
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Showing 1-2 of 2
Compare
Product Number
Description
Pricing