
L-α-Phosphatidylethanolamine from egg yolk

Synonym(s):
1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin
CAS Number:
EC Number:
Products
Product Category
Brand
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Optical Isomer
Showing 1-2 of 2