
DMT-2′Fluoro-dG(ib) Phosphoramidite

Synonym(s):
DMT-2′Fluoro-dG(ib) Amidite
Empirical Formula (Hill Notation):
C44H53FN7O8P
CAS Number:
Molecular Weight:
857.91
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing