
Diphenamid

Synonym(s):
N,N-Dimethyl-2,2-diphenylacetamide
Linear Formula:
(C6H5)2CHCON(CH3)2
CAS Number:
Molecular Weight:
239.31
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing