
Dihydrokainic acid

Synonym(s):
2-Carboxy-4-isopropyl-3-pyrrolidineacetic acid
Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing