
Chlorodiphenylsilane

Synonym(s):
Diphenylchlorosilane
Linear Formula:
(C6H5)2SiHCl
CAS Number:
Molecular Weight:
218.75
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing