![α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol Mr 2000](/deepweb/assets/sigmaaldrich/product/structures/784/232/8cbf268f-11e5-4d1b-aa37-5c62ae3e71f4/640/8cbf268f-11e5-4d1b-aa37-5c62ae3e71f4.png)
α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol
![α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol Mr 2000](/deepweb/assets/sigmaaldrich/product/structures/784/232/8cbf268f-11e5-4d1b-aa37-5c62ae3e71f4/640/8cbf268f-11e5-4d1b-aa37-5c62ae3e71f4.png)
Synonym(s):
O,O′-Bis[2-(succinylamino)ethyl]polyethylene glycol
Linear Formula:
HOOCCH2CH2CONH(CH2CH2O)nCH2CH2NHCOCH2CH2COOH
CAS Number:
Products
Brand
Physical Form
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing