
Amiloride for peak identification

Synonym(s):
Amiloride hydrochloride dihydrate, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
Empirical Formula (Hill Notation):
C6H8ClN7O · HCl · 2H2O
CAS Number:
Molecular Weight:
302.12
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing