
Aervent® 50 Capsule Filter

Synonym(s):
EPA Est. 041237-FRA-001
Products
Product Category
Brand
Product Line
Sterilization Compatibility
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Hydrophobicity
Housing Material
Membrane Material
Pore size (µm)
Sterility
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing