
3-(Methylthio)-1-hexanol

Linear Formula:
CH3CH2CH2CH(SCH3)CH2CH2OH
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing