Skip to Content
Merck
All Photos(2)

Key Documents

384100

Sigma-Aldrich

Ethyl viologen diperchlorate

98%

Synonym(s):

1,1′-Diethyl-4,4′-bipyridinium diperchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cl2N2O8
CAS Number:
Molecular Weight:
413.21
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

98%

form

solid

mp

270 °C (dec.) (lit.)

solubility

H2O: 2.5%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.CC[n+]1ccc(cc1)-c2cc[n+](CC)cc2

InChI

1S/C14H18N2.2ClHO4/c1-3-15-9-5-13(6-10-15)14-7-11-16(4-2)12-8-14;2*2-1(3,4)5/h5-12H,3-4H2,1-2H3;2*(H,2,3,4,5)/q+2;;/p-2

InChI key

HTNDMKDAPWJPEX-UHFFFAOYSA-L

General description

Ethyl viologen diperchlorate (EV (ClO4)2) in PEO matrix may be a good option for making viologen based eletrochromic device for testing purposes as it contains EV2+, that is a good electron acceptor and perchlorate ion which serves as electrolyte. It is used for fabricating electrochromic devices.

Application

Ethyl viologen diperchlorate has been used in in-situ spectroscopic studies of viologen based electrochromic device.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-situ spectroscopic studies of viologen based electrochromic device.
Mishra S, et al.
Opt. Mater., 66, 65-71 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service