Merck
  • Pannexin-1 mediates large pore formation and interleukin-1beta release by the ATP-gated P2X7 receptor.
Pannexin-1 mediates large pore formation and interleukin-1beta release by the ATP-gated P2X7 receptor.
The EMBO journal (2006-10-13)
Pablo Pelegrin, Annmarie Surprenant
ABSTRACT
P2X(7) receptors are ATP-gated cation channels; their activation in macrophage also leads to rapid opening of a membrane pore permeable to dyes such as ethidium, and to release of the pro-inflammatory cytokine, interleukin-1beta (IL-1beta). It has not been known what this dye-uptake path is, or whether it is involved in downstream signalling to IL-1beta release. Here, we identify pannexin-1, a recently described mammalian protein that functions as a hemichannel when ectopically expressed, as this dye-uptake pathway and show that signalling through pannexin-1 is required for processing of caspase-1 and release of mature IL-1beta induced by P2X(7) receptor activation.
Sigma-Aldrich
Scrambled 10Panx1 trifluoroacetate salt, ≥98% (HPLC)
Sigma-Aldrich
10Panx1 trifluoroacetate salt, ≥98% (HPLC)