Merck
  • A high mitochondrial transport rate characterizes CNS neurons with high axonal regeneration capacity.
A high mitochondrial transport rate characterizes CNS neurons with high axonal regeneration capacity.
PloS one (2017-09-20)
Romain Cartoni, Gulcin Pekkurnaz, Chen Wang, Thomas L Schwarz, Zhigang He
ABSTRACT
Improving axonal transport in the injured and diseased central nervous system has been proposed as a promising strategy to improve neuronal repair. However, the contribution of each cargo to the repair mechanism is unknown. DRG neurons globally increase axonal transport during regeneration. Because the transport of specific cargos after axonal insult has not been examined systematically in a model of enhanced regenerative capacity, it is unknown whether the transport of all cargos would be modulated equally in injured central nervous system neurons. Here, using a microfluidic culture system we compared neurons co-deleted for PTEN and SOCS3, an established model of high axonal regeneration capacity, to control neurons. We measured the axonal transport of three cargos (mitochondria, synaptic vesicles and late endosomes) in regenerating axons and found that the transport of mitochondria, but not the other cargos, was increased in PTEN/SOCS3 co-deleted axons relative to controls. The results reported here suggest a pivotal role for this organelle during axonal regeneration.
Sigma-Aldrich
Kynurenic acid, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Trypsin inhibitor from chicken egg white, Type II-O, Partially purified ovomucoid, containing ovoinhibitor
Pricing and availability is not currently available.