Merck
Systematic approach in Mg
Analytica chimica acta (2017-09-17)
L Lvova, C Guanais Gonçalves, L Prodi, M Sgarzi, N Zaccheroni, M Lombardo, A Legin, C Di Natale, R Paolesse
ABSTRACT
A systematic study of a series of diaza-18-crown-6 8-hydroxyquinoline (DCHQ) chemosensors, devoted to Mg(II) ion detection, was performed. Functionalization of DCHQ by peripheral substituents allowed the development of novel all-solid-state optodes via inclusion inside PVC-based polymeric films. The influence on the DCHQ-based optode response of the lipophilic sites functionalization and of the nature of the plasticizer, was investigated. Fluorimetric studies on optodes sensitivity towards a number of different metal cations (Ca
Product Description
Supelco
Magnesium ionophore VII, Selectophore, function tested
SKU
Pack Size
Availability
Price
Quantity
Supelco
Chromoionophore I, Selectophore