Merck
5,6-Di-2-thienyl-2,3-dihydro-pyrazine.
Acta crystallographica. Section E, Structure reports online (2010-01-01)
Madhukar Hemamalini, Hoong-Kun Fun
ABSTRACT
In the title compound, C(12)H(10)N(2)S(2), which was synthesized by the reaction of 2,2'-thenil and ethyl-enediamine, the dihedral angle between the two thio-phene rings is 66.33 (9)°. In the crystal structure, inter-molecular C-H⋯N hydrogen bonds link the mol-ecules into infinite chains along the b axis and weak C-H⋯π inter-actions may further stabilize the structure.
Sigma-Aldrich
2,2′-Thenil, 98%