Merck
  • Determination of γ-hydroxybutyrate (GHB), β-hydroxybutyrate (BHB), pregabalin, 1,4-butane-diol (1,4BD) and γ-butyrolactone (GBL) in whole blood and urine samples by UPLC-MSMS.
Determination of γ-hydroxybutyrate (GHB), β-hydroxybutyrate (BHB), pregabalin, 1,4-butane-diol (1,4BD) and γ-butyrolactone (GBL) in whole blood and urine samples by UPLC-MSMS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-01-10)
Sandra Rinne Dahl, Kirsten Midtbøen Olsen, Dag Helge Strand
ABSTRACT
The demand of high throughput methods for the determination of gamma-hydroxybutyrate (GHB) and its precursors gamma-butyrolactone (GBL) and 1,4-butane-diol (1,4BD) as well as for pregabalin is increasing. Here we present two analytical methods using ultra-high pressure liquid chromatography (UPLC) and tandem mass spectrometric (MS/MS) detection for the determination of GHB, beta-hydroxybutyrate (BHB), pregabalin, 1,4BD and GBL in whole blood and urine. Using the 96-well formate, the whole blood method is a simple high-throughput method suitable for screening of large sample amounts. With an easy sample preparation for urine including only dilution and filtration of the sample, the method is suitable for fast screening of urine samples. Both methods showed acceptable linearity, acceptable limits of detection, and limits of quantification. The within-day and between-day precisions of all analytes were lower than 10% RSD. The analytes were extracted from matrices with recoveries near 100%, and no major matrix effects were observed. Both methods have been used as routine screening analyses of whole blood and urine samples since January 2010.
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, ≥99%
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, 99%