Merck
  • Thermally stable AgI quantum-dot-based room-temperature fast ionic conductors.
Thermally stable AgI quantum-dot-based room-temperature fast ionic conductors.
Small (Weinheim an der Bergstrasse, Germany) (2011-11-01)
Biao Xu, Xun Wang
ABSTRACT
AgI quantum dot (QD)-based room-temperature fast ionic conductors are prepared via an aqueous route. New phase behavior and good performance are found in this material. This is a prototype of quantum dot-ionics.
Sigma-Aldrich
Silver iodide, 99%
Sigma-Aldrich
Silver iodide, 99.999% trace metals basis