Merck
  • Second harmonic generation in gallium phosphide photonic crystal nanocavities with ultralow continuous wave pump power.
Second harmonic generation in gallium phosphide photonic crystal nanocavities with ultralow continuous wave pump power.
Optics express (2010-01-07)
Kelley Rivoire, Ziliang Lin, Fariba Hatami, W Ted Masselink, Jelena Vucković
ABSTRACT
We demonstrate second harmonic generation in photonic crystal nanocavities fabricated in the semiconductor gallium phosphide. We observe second harmonic radiation at 750 nm with input powers of only nanowatts coupled to the cavity and conversion effciency P(out)/P(2)(in,coupled)=430%/W. The large electronic band gap of GaP minimizes absorption loss, allowing effcient conversion. Our results are promising for integrated, low-power light sources and on-chip reduction of input power in other nonlinear processes.
Sigma-Aldrich
Gallium phosphide, (single crystal substrate), <111>, diam. × thickness 2 in. × 0.5 mm