Merck
Pool chemical blast injury.
Annals of emergency medicine (2009-10-27)
Brian W Shippert
ABSTRACT
Swimming pools are one of the most popular forms of recreation in the United States. Pool-related injuries may produce significant morbidity and mortality, and those related to pool chemicals are of particular importance. The majority of injuries associated with pool chemicals are respiratory, with the remainder composed mainly of dermal exposures. There are few case reports about injuries from pool chemicals, and the potential for significant injury from blast force is presented here.
Sigma-Aldrich
Calcium hypochlorite, technical grade
Sigma-Aldrich
Calcium hypochlorite, granular, SAJ first grade, available chlorine ≥60 %