Merck
  • Biosorption of Cr(III) from aqueous solution using algal biomass spirogyra spp.
Biosorption of Cr(III) from aqueous solution using algal biomass spirogyra spp.
Journal of hazardous materials (2006-12-07)
Narsi R Bishnoi, Rajender Kumar, Sunil Kumar, Suman Rani
ABSTRACT
In the present investigation, a fresh water green algae spirogyra spp. was used as an inexpensive and efficient biosorbent for Cr(III) removal from aqueous solution. The algal biomass was treated with 0.1M NaOH, 0.2M CaCl(2) and 5% HCHO. The biosorption efficiency was compared with untreated biomass. The effects of various physico-chemical parameters were studied, e.g. pH 3.0-6.0, initial metal ions concentration 20-150mgL(-1), algal dose 1.0-3.0gL(-1), and contact time 15-180min, respectively. Biosorption of Cr(III) is highly pH dependent. Maximum 81.02% adsorption of Cr(III) was observed with 0.2M CaCl(2) treated biomass at pH 5.0. Removal of Cr(III) was more than 70% in 45min of contact time with different treated and untreated algal biomass at concentration 30mgL(-1). Maximum metal uptake (Q(max)) was observed as 30.21mgg(-1) with 0.2M CaCl(2) treated algal biomass indicate good biosorbents than other treated and untreated biomass. The high values of correlation coefficient (r(2)<0.90) indicate equilibrium data of treated and untreated form of algal biomass well fitted in Freundlich than Langmuir isotherms model equations.
Sigma-Aldrich
Chromium(III) chloride, 99.99% trace metals basis
Sigma-Aldrich
Chromium(III) chloride, purified by sublimation, 99%