Merck
  • Repetitive mRNA vaccination is required to improve the quality of broad-spectrum anti-SARS-CoV-2 antibodies in the absence of CXCL13.
Repetitive mRNA vaccination is required to improve the quality of broad-spectrum anti-SARS-CoV-2 antibodies in the absence of CXCL13.
Science advances (2023-08-04)
Marne Azarias Da Silva, Pierre Nioche, Calaiselvy Soudaramourty, Anne Bull-Maurer, Mounira Tiouajni, Dechuan Kong, Ouafa Zghidi-Abouzid, Morgane Picard, Ana Mendes-Frias, André Santa-Cruz, Alexandre Carvalho, Carlos Capela, Jorge Pedrosa, António Gil Castro, Paul Loubet, Albert Sotto, Laurent Muller, Jean-Yves Lefrant, Claire Roger, Pierre-Géraud Claret, Sandra Duvnjak, Tu-Anh Tran, Kenzo Tokunaga, Ricardo Silvestre, Pierre Corbeau, Fabrizio Mammano, Jérôme Estaquier
ABSTRACT
Since the initial spread of severe acute respiratory syndrome coronavirus 2 infection, several viral variants have emerged and represent a major challenge for immune control, particularly in the context of vaccination. We evaluated the quantity, quality, and persistence of immunoglobulin G (IgG) and IgA in individuals who received two or three doses of messenger RNA (mRNA) vaccines, compared with previously infected vaccinated individuals. We show that three doses of mRNA vaccine were required to match the humoral responses of preinfected vaccinees. Given the importance of antibody-dependent cell-mediated immunity against viral infections, we also measured the capacity of IgG to recognize spike variants expressed on the cell surface and found that cross-reactivity was also strongly improved by repeated vaccination. Last, we report low levels of CXCL13, a surrogate marker of germinal center activation and formation, in vaccinees both after two and three doses compared with preinfected individuals, providing a potential explanation for the short duration and low quality of Ig induced.
Sigma-Aldrich
Anti-Human IgA (α-chain specific), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody