Merck
Validation of the biotinyl ligand-avidin-gold technique.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1992-05-01)
R E Morris, G M Ciraolo, C B Saelinger
ABSTRACT
We demonstrate here that the intracellular routing of biotinylated ligands was not affected by the attachment of streptavidin gold colloids so long as the electron-dense marker was added after the biotinyl ligand-receptor interaction had occurred. The binding, internalization, and intracellular routing of three different biotinyl ligands were followed in mouse LM fibroblasts. The biotinyl (B) ligands included B-choleragenoid (B-CTd), B-wheat germ agglutinin (B-WGA), and B-Pseudomonas exotoxin A (B-PE). All three ligands showed distinct intracellular trafficking patterns. B-WGA and B-PE entered via clathrin-coated pits, whereas B-CTd did not. After entry, B-CTd was routed to the lysosomal compartment without involvement of the Golgi. Although B-PE and B-WGA were also routed to the lysosomal compartment, a significant portion of these two ligands was observed in association with the Golgi. B-WGA, however, remained in the endosomal and Golgi compartments longer than did B-PE. We also monitored the internalization and routing of native PE by an indirect immunoperoxidase technique done in conjunction with saponin solubilization. The results corroborated the observations with the biotinyl-PE-streptavidin-gold method. In contrast, biotinyl-PE added to streptavidin-gold before addition to LM cells was poorly internalized and routed aberrantly. From these observations we conclude that the biotinyl ligand-avidin-gold technique is a valid method for following the binding, internalization, and intracellular routing of ligands.
Sigma-Aldrich
Lectin from Triticum vulgaris (wheat), biotin conjugate, lyophilized powder