Merck
  • GPR126 regulates colorectal cancer cell proliferation by mediating HDAC2 and GLI2 expression.
GPR126 regulates colorectal cancer cell proliferation by mediating HDAC2 and GLI2 expression.
Cancer science (2021-02-26)
Hengxiang Cui, Wenjie Yu, Minhao Yu, Yang Luo, Mingming Yang, Ruochen Cong, Xin Chu, Ganglong Gao, Ming Zhong
ABSTRACT
The G-protein-coupled receptor 126 (GPR126) may play an important role in tumor development, although its role remains poorly understood. We found that GPR126 had higher expression in most colorectal cancer cell lines than in normal colon epithelial cell lines, and higher expression levels in colorectal cancer tissues than in normal adjacent colon tissues. GPR126 knockdown induced by shRNA inhibited cell viability and colony formation in HT-29, HCT116, and LoVo cells, decreased BrdU incorporation into newly synthesized proliferating HT-29 cells, led to an arrest of cell cycle progression at the G1 phase in HCT-116 and HT-29 cells, and suppressed tumorigenesis of HT-29, HCT116, and LoVo cells in nude mouse xenograft models. GPR126 knockdown engendered decreased transcription and translation of histone deacetylase 2 (HDAC2), previously implicated in the activation of GLI1 and GLI2 in the Hedgehog signaling pathway. Ectopic expression of HDAC2 in GPR126-silenced cells restored cell viability and proliferation, GLI2 luciferase reporter activity, partially recovered GLI2 expression, and reduced the cell cycle arrest. HDAC2 regulated GLI2 expression and, along with GLI2, it bound to the PTCH1 promoter, as evidenced by a chip assay with HT-29 cells. Purmorphamine, a hedgehog agonist, largely restored the cell viability and expression of GLI2 proteins in GPR126-silenced HT-29 cells, whereas GANT61, a hedgehog inhibitor, further enhanced the GPR126 knockdown-induced inhibitory effects. Our findings demonstrate that GPR126 regulates colorectal cancer cell proliferation by mediating the expression of HDAC2 and GLI2, therefore it may represent a suitable therapeutic target for colorectal cancer treatment.
Sigma-Aldrich
Anti-Histone Deacetylase 1 (HDAC1) antibody, Mouse monoclonal, clone HDAC1-21, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Histone Deacetylase 2 (HDAC2) antibody, Mouse monoclonal, clone HDAC2-62, purified from hybridoma cell culture