Merck
  • Moderate prenatal alcohol exposure increases total length of L1-expressing axons in E15.5 mice.
Moderate prenatal alcohol exposure increases total length of L1-expressing axons in E15.5 mice.
Neurotoxicology and teratology (2021-02-27)
Avery Sicher, Shannon Kiss, Paige Springmann, Karen Herrera, Abigail McElroy, Kelsey Blake, Emily Crocker, Christa Jacob, McKayla Lefkove, Myla Cramer, Allysen Henriksen, Josef Novacek, Jenna Severa, Justin Siberski, Emily Thomas, Peter Chi, Carlita Favero
ABSTRACT
Public health campaigns broadcast the link between heavy alcohol consumption during pregnancy and physical, cognitive, and behavioral birth defects; however, they appear less effective in deterring moderate consumption prevalent in women who are pregnant or of childbearing age. The incidence of mild Fetal Alcohol Spectrum Disorders (FASD) is likely underestimated because the affected individuals lack physical signs such as retarded growth and facial dysmorphology and cognitive/behavioral deficits are not commonly detected until late childhood. Sensory information processing is distorted in FASD, but alcohol's effects on the development of axons that mediate these functions are not widely investigated. We hypothesize that alcohol exposure alters axon growth and guidance contributing to the aberrant connectivity that is a hallmark of FASD. To test this, we administered alcohol to pregnant dams from embryonic day (E) 7.5 to 14.5, during the time that axons which form the major forebrain tracts are growing. We found that moderate alcohol exposure had no effect on body weight of E15.5 embryos, but significantly increased the length of L1+ axons. To investigate a possible cause of increased L1+ axon length, we investigated the number and distribution of corridor cells, one of multiple guidance cues for thalamocortical axons which are involved in sensory processing. Alcohol did not affect corridor cell number or distribution at the time when thalamocortical axons are migrating. Future studies will investigate the function of other guidance cues for thalamocortical axons, as well as lasting consequences of axon misguidance with prenatal alcohol exposure.
Sigma-Aldrich
JumpStart REDTaq® ReadyMix Reaction Mix, for PCR
Pricing and availability is not currently available.