Merck
  • Cytokine Kinetics in the First Week of Tuberculosis Therapy as a Tool to Confirm a Clinical Diagnosis and Guide Therapy.
Cytokine Kinetics in the First Week of Tuberculosis Therapy as a Tool to Confirm a Clinical Diagnosis and Guide Therapy.
PloS one (2015-06-27)
Alice L den Hertog, María Montero-Martín, Rachel L Saunders, Matthew Blakiston, Sandra Menting, Jeevan B Sherchand, Lovett Lawson, Olanrewaju Oladimeji, Saddiq T Abdurrahman, Luis E Cuevas, Richard M Anthony
ABSTRACT
Many patients treated for tuberculosis (TB) in low and middle income countries are treated based on clinical suspicion without bacteriological confirmation. This is often due to lack of rapid simple accurate diagnostics and low healthcare provider confidence in the predictive value of current tests. We previously reported in an animal TB model that levels of host markers rapidly change in response to treatment initiation. We assessed the potential of host biomarker kinetics of TB patients during the first two weeks of therapy to identify patients responding to treatment. Adult patients clinically diagnosed with and treated for TB, 29 in Nigeria and 24 in Nepal, were analyzed. Changes in concentrations of non-specific host biomarkers, particularly IP-10, in response to the first week of anti-TB therapy were strongly associated with bacteriological confirmation of TB. A decrease in IP-10 level of >300pg/ml between 0 and 7 days of treatment identified 75% of both smear-positive and smear-negative culture positive patients and correctly excluded TB in all nine culture negative patients. Monitoring of early IP-10 responses to treatment could form the basis of a simplified assay and could help identify patients who were erroneously clinically diagnosed with TB or those infected with drug resistant strains on inappropriate treatment. We believe this approach may be particularly appropriate for difficult to diagnose patients, e.g. smear-negative HIV-positive, or those with extra-pulmonary TB, often treated without bacterial confirmation.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel III - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.