Merck
  • Hsp70 protects against UVB induced apoptosis by preventing release of cathepsins and cytochrome c in human melanocytes.
Hsp70 protects against UVB induced apoptosis by preventing release of cathepsins and cytochrome c in human melanocytes.
Carcinogenesis (2006-09-05)
Cecilia Bivik, Inger Rosdahl, Karin Ollinger
ABSTRACT
Stress-induced heat shock protein 70 (Hsp70) effectively protects cells against apoptosis, although the anti-apoptotic mechanism is still undefined. Exposure of human melanocytes to heat and subsequent UVB irradiation increased the level of Hsp70 and pre-heating reduced UVB induced apoptosis. Immunofluorescence staining of Hsp70 in combination with staining of lysosomes (Lamp2) or mitochondria (Mitotracker) in pre-heated UVB exposed cells showed co-localization of Hsp70 with both lysosomes and mitochondria in the surviving cell population. Furthermore, UVB induced apoptosis was accompanied by lysosomal and mitochondrial membrane permeabilization, detected as release of cathepsin D and cytochrome c, respectively, which were prevented by heat pre-treatment. In purified fractions of lysosomes and mitochondria, recombinant Hsp70 attached to both lysosomal and mitochondrial membranes. Moreover, in apoptotic cells Bax was translocated from a diffuse cytosolic location into punctate mitochondrial-like structures, which was inhibited by Hsp70 induction. Such inhibition of Bax translocation was abolished by transfection with Hsp70 siRNA. Furthermore, Hsp70 siRNA eliminated the apoptosis preventive effect observed after pre-heating. These findings show Hsp70 to rescue melanocytes from UVB induced apoptosis by preventing release of cathepsins from lysosomes, Bax translocation and cytochrome c release from mitochondria.
Sigma-Aldrich
Anti-Cathepsin D (Ab-1) Mouse mAb (BC011), liquid, clone BC011, Calbiochem®
Pricing and availability is not currently available. Contact Technical Service