Merck
  • The impact of capsaicinoids on APP processing in Alzheimer's disease in SH-SY5Y cells.
The impact of capsaicinoids on APP processing in Alzheimer's disease in SH-SY5Y cells.
Scientific reports (2020-06-10)
Marcus O W Grimm, Tamara Blümel, Anna A Lauer, Daniel Janitschke, Christoph Stahlmann, Janine Mett, Viola J Haupenthal, Anna-Maria Miederer, Barbara A Niemeyer, Heike S Grimm, Tobias Hartmann
ABSTRACT
The vanilloid capsaicin is a widely consumed spice, known for its burning and "hot" sensation through activation of TRPV1 ion-channels, but also known to decrease oxidative stress, inflammation and influence tau-pathology. Beside these positive effects, little is known about its effects on amyloid-precursor-protein (APP) processing leading to amyloid-β (Aβ), the major component of senile plaques. Treatment of neuroblastoma cells with capsaicinoids (24 hours, 10 µM) resulted in enhanced Aβ-production and reduced Aβ-degradation, leading to increased Aβ-levels. In detailed analysis of the amyloidogenic-pathway, both BACE1 gene-expression as well as protein-levels were found to be elevated, leading to increased β-secretase-activity. Additionally, γ-secretase gene-expression as well as activity was enhanced, accompanied by a shift of presenilin from non-raft to raft membrane-domains where amyloidogenic processing takes place. Furthermore, impaired Aβ-degradation in presence of capsaicinoids is dependent on the insulin-degrading-enzyme, one of the major Aβ-degrading-enzymes. Regarding Aβ-homeostasis, no differences were found between the major capsaicinoids, capsaicin and dihydrocapsaicin, and a mixture of naturally derived capsaicinoids; effects on Ca2+-homeostasis were ruled out. Our results show that in respect to Alzheimer's disease, besides the known positive effects of capsaicinoids, pro-amyloidogenic properties also exist, enhancing Aβ-levels, likely restricting the potential use of capsaicinoids as therapeutic substances in Alzheimer's disease.
Sigma-Aldrich
γ-Secretase Inhibitor IV, The γ-Secretase Inhibitor IV controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.
Sigma-Aldrich
Anti-BACE 1, C-Terminus (485-501) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
β-Secretase Inhibitor II, The β-Secretase Inhibitor II, also referenced under CAS 263563-09-3, controls the biological activity of β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.