Merck
  • Scalable neutral H2O2 electrosynthesis by platinum diphosphide nanocrystals by regulating oxygen reduction reaction pathways.
Scalable neutral H2O2 electrosynthesis by platinum diphosphide nanocrystals by regulating oxygen reduction reaction pathways.
Nature communications (2020-08-09)
Hui Li, Peng Wen, Dominique S Itanze, Zachary D Hood, Shiba Adhikari, Chang Lu, Xiao Ma, Chaochao Dun, Lin Jiang, David L Carroll, Yejun Qiu, Scott M Geyer
ABSTRACT
Despite progress in small scale electrocatalytic production of hydrogen peroxide (H2O2) using a rotating ring-disk electrode, further work is needed to develop a non-toxic, selective, and stable O2-to-H2O2 electrocatalyst for realizing continuous on-site production of neutral hydrogen peroxide. We report ultrasmall and monodisperse colloidal PtP2 nanocrystals that achieve H2O2 production at near zero-overpotential with near unity H2O2 selectivity at 0.27 V vs. RHE. Density functional theory calculations indicate that P promotes hydrogenation of OOH* to H2O2 by weakening the Pt-OOH* bond and suppressing the dissociative OOH* to O* pathway. Atomic layer deposition of Al2O3 prevents NC aggregation and enables application in a polymer electrolyte membrane fuel cell (PEMFC) with a maximum r(H2O2) of 2.26 mmol h-1 cm-2 and a current efficiency of 78.8% even at a high current density of 150 mA cm-2. Catalyst stability enables an accumulated neutral H2O2 concentration in 600 mL of 3.0 wt% (pH = 6.6).
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Tris(trimethylsilyl)phosphine, 95%