Merck
  • The CK2 phosphorylation of vitronectin. Promotion of cell adhesion via the alpha(v)beta 3-phosphatidylinositol 3-kinase pathway.
The CK2 phosphorylation of vitronectin. Promotion of cell adhesion via the alpha(v)beta 3-phosphatidylinositol 3-kinase pathway.
The Journal of biological chemistry (2001-03-30)
D Seger, R Seger, S Shaltiel
ABSTRACT
Phosphorylation of vitronectin (Vn) by casein kinase II was previously shown to occur at Thr50 and Thr57 and to augment a major physiological function of vitronectin-cell adhesion and spreading. Here we show that this phosphorylation increases cell adhesion via the alpha(v)beta3 (not via the alpha(v)beta5 integrin), suggesting that alpha(v)beta3 differs from alpha(v)beta5 in its biorecognition profile. Although both the phospho (CK2-PVn) and non-phospho (Vn) analogs of vitronectin (simulated by mutants Vn(T50E,T57E), and Vn(T50A,T57A), respectively) trigger the alpha(v)beta3 as well as the alpha(v)beta5 integrins, and equally activate the ERK pathway, these two forms are different in their activation of the focal adhesion kinase/phosphatidylinositol 3-kinase (PI3K)/protein kinase B (PKB) pathway. Specifically, we show (i) that, upon exposure of cells to Vn/CK2-PVn, their PKB activation depends on the availability of the alpha(v)beta3 integrin on their surface; (ii) that upon adhesion of the beta3-transfected cells onto the CK2-PVn, the extent of PKB activation coincides with the enhanced adhesion of these cells, and (iii) that both the PKB activation and the elevation in the adhesion of these cells is PI3K-dependent. The occurrence of a cell surface receptor that specifically distinguishes between a phosphorylated and a non-phosphorylated analog of Vn, together with the fact that it preferentially activates a distinct intra-cellular signaling pathway, suggest that extra-cellular CK2 phosphorylation may play an important role in the regulation of cell adhesion and migration.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone BB10, clone BB10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse