Merck
  • Innate and adaptive nasal mucosal immune responses following experimental human pneumococcal colonization.
Innate and adaptive nasal mucosal immune responses following experimental human pneumococcal colonization.
The Journal of clinical investigation (2019-07-31)
Simon P Jochems, Karin de Ruiter, Carla Solórzano, Astrid Voskamp, Elena Mitsi, Elissavet Nikolaou, Beatriz F Carniel, Sherin Pojar, Esther L German, Jesús Reiné, Alessandra Soares-Schanoski, Helen Hill, Rachel Robinson, Angela D Hyder-Wright, Caroline M Weight, Pascal F Durrenberger, Robert S Heyderman, Stephen B Gordon, Hermelijn H Smits, Britta C Urban, Jamie Rylance, Andrea M Collins, Mark D Wilkie, Lepa Lazarova, Samuel C Leong, Maria Yazdanbakhsh, Daniela M Ferreira
ABSTRACT
Streptococcus pneumoniae (Spn) is a common cause of respiratory infection, but also frequently colonizes the nasopharynx in the absence of disease. We used mass cytometry to study immune cells from nasal biopsy samples collected following experimental human pneumococcal challenge in order to identify immunological mechanisms of control of Spn colonization. Using 37 markers, we characterized 293 nasal immune cell clusters, of which 7 were associated with Spn colonization. B cell and CD8+CD161+ T cell clusters were significantly lower in colonized than in non-colonized subjects. By following a second cohort before and after pneumococcal challenge we observed that B cells were depleted from the nasal mucosa upon Spn colonization. This associated with an expansion of Spn polysaccharide-specific and total plasmablasts in blood. Moreover, increased responses of blood mucosal associated invariant T (MAIT) cells against in vitro stimulation with pneumococcus prior to challenge associated with protection against establishment of Spn colonization and with increased mucosal MAIT cell populations. These results implicate MAIT cells in the protection against pneumococcal colonization and demonstrate that colonization affects mucosal and circulating B cell populations.
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgA (α-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution