Merck
  • Molecularly Distinct Clathrin-Coated Pits Differentially Impact EGFR Fate and Signaling.
Molecularly Distinct Clathrin-Coated Pits Differentially Impact EGFR Fate and Signaling.
Cell reports (2019-06-06)
Roberta Pascolutti, Veronica Algisi, Alexia Conte, Andrea Raimondi, Mithun Pasham, Srigokul Upadhyayula, Raphael Gaudin, Tanja Maritzen, Elisa Barbieri, Giusi Caldieri, Chiara Tordonato, Stefano Confalonieri, Stefano Freddi, Maria Grazia Malabarba, Elena Maspero, Simona Polo, Carlo Tacchetti, Volker Haucke, Tom Kirchhausen, Pier Paolo Di Fiore, Sara Sigismund
ABSTRACT
Adaptor protein 2 (AP2) is a major constituent of clathrin-coated pits (CCPs). Whether it is essential for all forms of clathrin-mediated endocytosis (CME) in mammalian cells is an open issue. Here, we demonstrate, by live TIRF microscopy, the existence of a subclass of relatively short-lived CCPs lacking AP2 under physiological, unperturbed conditions. This subclass is retained in AP2-knockout cells and is able to support the internalization of epidermal growth factor receptor (EGFR) but not of transferrin receptor (TfR). The AP2-independent internalization mechanism relies on the endocytic adaptors eps15, eps15L1, and epsin1. The absence of AP2 impairs the recycling of the EGFR to the cell surface, thereby augmenting its degradation. Accordingly, under conditions of AP2 ablation, we detected dampening of EGFR-dependent AKT signaling and cell migration, arguing that distinct classes of CCPs could provide specialized functions in regulating EGFR recycling and signaling.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, 351-368) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Anti-ZAP-70 Antibody, clone 2F3.2, FITC conjugate, clone 2F3.2, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Adaptin antibody produced in mouse, clone 100/2, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution