Merck
Type B Insulin Resistance Masquerading as Ovarian Hyperthecosis.
The Journal of clinical endocrinology and metabolism (2016-12-03)
Rebecca J Brown, Jalaja Joseph, Elaine Cochran, Cornelia Gewert, Robert Semple, Phillip Gorden
ABSTRACT
Hyperinsulinemia can lead to pathologic ovarian growth and androgen production. A 29-year-old woman developed an autoantibody to the insulin receptor (type B insulin resistance), causing extreme insulin resistance and hyperinsulinemia. Testosterone levels were elevated to the adult male range. Treatment with gonadotropin-releasing hormone (GnRH) analog led to normalization of testosterone, despite persistent extreme insulin resistance. This case demonstrates that gonadotropins are necessary for insulin to cause pathologic ovarian androgen production. Suppression of gonadotropins with GnRH analogs may be a useful therapeutic option in patients with severe hyperandrogenism or ovarian enlargement because of hyperinsulinemia.
Sigma-Aldrich
Anti-Human IgG (Fc Specific)āˆ’Agarose antibody produced in goat, affinity isolated antibody, PBS suspension