
tert-Butyl 4-(3-(hydroxymethyl)phenyl)piperidine-1-carboxylate

Empirical Formula (Hill Notation):
C17H25NO3
CAS Number:
Molecular Weight:
291.39
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing