
Palonosetron Related Compound C

Synonym(s):
(3aS)-2-[(3R)-1-Azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benz[de]isoquinolin-1-one monohydrochloride, (3aS)-2-[(3R)-Quinuclidin-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride
Empirical Formula (Hill Notation):
C19H24N2O · HCl
CAS Number:
Molecular Weight:
332.87
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2