
N-Nitrosomethylaminobutyric Acid (NMBA)

Synonym(s):
4-[Methyl(nitroso)amino]butanoic acid
Empirical Formula (Hill Notation):
C5H10N2O3
CAS Number:
Molecular Weight:
146.14
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2