
Monoclonal Anti-Tumor Necrosis Factor-α–Fluorescein antibody produced in mouse

Synonym(s):
Monoclonal Anti-Tumor Necrosis Factor-a-Carboxyfluorescein, Anti-TNF-α
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing