Skip to Content
Merck
Methyl δ-aminolevulinate hydrochloride ≥98% (TLC)

Methyl δ-aminolevulinate hydrochloride

Methyl δ-aminolevulinate hydrochloride ≥98% (TLC)
Synonym(s):
5-Amino-4-oxopentanoic acid methyl ester, 5-Aminolevulinic acid methyl ester
Empirical Formula (Hill Notation):
C6H11NO3 · HCl
CAS Number:
Molecular Weight:
181.62
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing