
Marrubiin

Synonym(s):
Marrubin
Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing