
MAN-5 Glycan

Synonym(s):
(Man)5(GlcNAc)2, Mannopentaose-di-(N-acetyl-D-glucosamine), Oligomannose-5 glycan
Empirical Formula (Hill Notation):
C46H78N2O36
CAS Number:
Molecular Weight:
1235.10
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing