
Lutetium

Synonym(s):
Lutetium
Empirical Formula (Hill Notation):
Lu
CAS Number:
Molecular Weight:
174.97
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Element
Showing 1-9 of 9
Compare
Product Number
Description
Pricing