
Lactulose for peak identification

Synonym(s):
Lactulose, 4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Products
Formula Weight
Manufacturer
Format
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing