
Formaldehyde neutral buffer solution

Synonym(s):
Formaldehyde solution, Formalin
Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing