
Fluconazole Impurity F

Synonym(s):
(2RS)-2-(2,4-Difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propane-1,2-diol
Empirical Formula (Hill Notation):
C11H11F2N3O2
CAS Number:
Molecular Weight:
255.22
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing