
Dibutyl adipate

Synonym(s):
Adipic acid dibutyl ester
Linear Formula:
[CH2CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
258.35
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing