
Cholesteryl caprylate

Synonym(s):
3β-Hydroxy-5-cholestene 3-octanoate, 5-Cholesten-3β-ol 3-octanoate, Cholesteryl n-octanoate, Cholesteryl octanoate
Empirical Formula (Hill Notation):
C35H60O2
CAS Number:
Molecular Weight:
512.85
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing