Skip to Content
Merck
All Photos(1)

Key Documents

RA4624-EP

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

A-4-62, for 5810/5810R, aerosol-tight, with 4 MTP buckets

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

to hold MTP, DWP, PCR plates
aerosol-tight
economic swing- bucket rotor
for 5810/5810R
with 4 MTP buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022638041 (NA)
Eppendorf® 5810711002 (EU)

parameter

2,750 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

  • A versatile swing-bucket rotor (A-4-62-MTP) accommodates 0.2 mL to 250 mL tubes and bottles as well as plates.
  • Buckets, lid and adapters are autoclavable at 121 °C, 20 min.
  • Aerosol-tight caps are available for the safe centrifugation of hazardous samples.
  • Rotor lifetime: max. 100,000 cycles.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service