Skip to Content
Merck
All Photos(1)

Key Documents

PZ0259

Sigma-Aldrich

PH-797804

≥98% (HPLC)

Synonym(s):

3-Bromo-4-[(2,4-difluorobenzyl)oxy]-1-[5-[(methylamino)carbonyl]-2-methylphenyl]-6-methylpyridin-2(1H)-one, 3-[3-Bromo-4-[(2,4-difluorophenyl)methoxy]-6-methyl-2-oxo-1(2H)-pyridinyl]-N,4-dimethyl-benzamide, PH 797804

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19BrF2N2O3
CAS Number:
Molecular Weight:
477.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

20-25°C

SMILES string

CC1=CC(OCC2=C(F)C=C(F)C=C2)=C(Br)C(N1C3=CC(C(NC)=O)=CC=C3C)=O

InChI

1S/C22H19BrF2N2O3/c1-12-4-5-14(21(28)26-3)9-18(12)27-13(2)8-19(20(23)22(27)29)30-11-15-6-7-16(24)10-17(15)25/h4-10H,11H2,1-3H3,(H,26,28)

InChI key

KCAJXIDMCNPGHZ-UHFFFAOYSA-N

Biochem/physiol Actions

PH-797804 is an orally active, potent and readily reversible ATP competitive inhibitor of the alpha isoform of human p38 MAP kinase. PH-797804 blocks inflammation-induced production of cytokines and proinflammatory mediators.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service